Stampa…

Anti-TLK2 Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT8040-10A-BIOTEA 1200 CHF
USBIT8040-10A-BIOT
Anti-TLK2 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TLK2 (tousled-like kinase 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR