Stampa…

Anti-TOB2 Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT7948-20A-BIOTEA 1200 CHF
USBIT7948-20A-BIOT
Anti-TOB2 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TOB2 (transducer of ERBB2, 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR