Stampa…

Anti-TMEM205 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT6020-75EA 774 CHF
USBIT6020-75
Anti-TMEM205 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: TMEM205 (transmembrane protein 205)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR