Stampa…

Anti-RNF139 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT5890EA 774 CHF
USBIT5890
Anti-RNF139 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:16000; Western Blot: 0.5-1. 5 ?g/ml

Type: Primary
Antigen: RNF139 (ring finger protein 139)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR