Stampa…

Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5600-12EA 818 CHF
USBIT5600-12
Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR