Stampa…

Anti-Tissue Plasminogen Activator Sheep Polyclonal Antibody

Fornitore: United States Biological
USBIT5600-05JEA 929 CHF
USBIT5600-05J
Anti-Tissue Plasminogen Activator Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR