Stampa…

Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIT5600-05EEA 1270 CHF
USBIT5600-05E
Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:300

Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR