Stampa…

Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT5600-04AEA 1510 CHF
USBIT5600-04A
Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: Western Blot: 5 ?g/ml

Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR