Stampa…
Anti-TIMP4 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT5587-02BEA 676 CHF
USBIT5587-02B
Anti-TIMP4 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot:1-2?g/ml
Type: Primary
Antigen: TIMP4 (TIMP metallopeptidase inhibitor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Mehr About VWR