Stampa…
Anti-Tissue Inhibitor of Metalloproteinase 2 Sheep Polyclonal Antibody
Fornitore: United States Biological
USBIT5585-10DEA 672 CHF
USBIT5585-10D
Anti-Tissue Inhibitor of Metalloproteinase 2 Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Tissue Inhibitor of Metalloproteinase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Mehr About VWR