Stampa…

Anti-IFRD1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5565-50EA 712 CHF
USBIT5565-50
Anti-IFRD1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-4?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: IFRD1 (interferon-related developmental regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Macaque, Mouse, Pig
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR