Stampa…

Anti-TP53INP2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: United States Biological
USBIT5501-20-APCEA 1200 CHF
USBIT5501-20-APC
Anti-TP53INP2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Flow Cytometry: 1:10-1:50; Immunochemistry: 1:50-1:100; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: TP53INP2 (tumor protein p53 inducible nuclear protein 2)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR