Stampa…

Anti-TIRAP Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5500-04EA 1060 CHF
USBIT5500-04
Anti-TIRAP Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml; Western Blot: 4?g/ml

Type: Primary
Antigen: TIRAP (toll-interleukin 1 receptor (TIR) domain containing adaptor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR