Stampa…

Anti-TINF2 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT5475-01BEA 889 CHF
USBIT5475-01B
Anti-TINF2 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 3. 75-7. 5 ?g/ml

Type: Primary
Antigen: TINF2 (TERF1 (TRF1)-interacting nuclear factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR