Stampa…

Anti-TIMELESS Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5470-03-100EA 966 CHF
USBIT5470-03-100
Anti-TIMELESS Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TIMELESS (timeless circadian clock)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR