Stampa…

Anti-HAVCR2 Rat Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n191]

Fornitore: United States Biological
USBIT5469-92REA 1050 CHF
USBIT5469-92R
Anti-HAVCR2 Rat Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n191]
Anticorpi
Type: Primary
Antigen: HAVCR2 (hepatitis A virus cellular receptor 2)
Clonality: monoclonal
Clone: 11n191
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR