Stampa…

Anti-HAVCR2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9L856]

Fornitore: United States Biological
USBIT5469-92FEA 1100 CHF
USBIT5469-92F
Anti-HAVCR2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9L856]
Anticorpi
Do not freeze!

Type: Primary
Antigen: HAVCR2 (hepatitis A virus cellular receptor 2)
Clonality: monoclonal
Clone: 9L856
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR