Stampa…

Anti-TIAL1/TIA Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT5461-95AEA 774 CHF
USBIT5461-95A
Anti-TIAL1/TIA Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:64000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: TIAL1/TIA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR