Stampa…

Anti-TRHR Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5430-20FEA 889 CHF
USBIT5430-20F
Anti-TRHR Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: TRHR (thyrotropin-releasing hormone receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR