Stampa…

Anti-Thyroid Transcription Factor 1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5405-01BEA 844 CHF
USBIT5405-01B
Anti-Thyroid Transcription Factor 1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunoblot: A 1:500-1:1000

Type: Primary
Antigen: Thyroid Transcription Factor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR