Stampa…

Anti-Thyroid Stimulating Hormone Mouse Monoclonal Antibody [clone: 3F366]

Fornitore: United States Biological
USBIT5400-49-250EA 462 CHF
USBIT5400-49-250
Anti-Thyroid Stimulating Hormone Mouse Monoclonal Antibody [clone: 3F366]
Anticorpi
Recommended dilution: Immunochemistry: 1:200-1:400

Type: Primary
Antigen: Thyroid Stimulating Hormone
Clonality: monoclonal
Clone: 3F366
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR