Stampa…

Anti-Thyroid Stimulating Hormone Mouse Monoclonal Antibody [clone: 9L711]

Fornitore: United States Biological
USBIT5400-13FEA 891 CHF
USBIT5400-13F
Anti-Thyroid Stimulating Hormone Mouse Monoclonal Antibody [clone: 9L711]
Anticorpi
Type: Primary
Antigen: Thyroid Stimulating Hormone
Clonality: monoclonal
Clone: 9L711
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR