Stampa…

Anti-Thyroid Stimulating Hormone, intact Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT5400-02DEA 490 CHF
USBIT5400-02D
Anti-Thyroid Stimulating Hormone, intact Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Thyroid Stimulating Hormone, intact
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR