Stampa…

Anti-TPO Mouse Monoclonal Antibody [clone: 11H102]

Fornitore: United States Biological
USBIT5399-01EEA 1100 CHF
USBIT5399-01E
Anti-TPO Mouse Monoclonal Antibody [clone: 11H102]
Anticorpi
Type: Primary
Antigen: TPO (thyroid peroxidase)
Clonality: monoclonal
Clone: 11H102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR