Stampa…

Anti-THRB Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT5349-01KEA 774 CHF
USBIT5349-01K
Anti-THRB Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: THRB (thyroid hormone receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR