Stampa…

Anti-TG Mouse Monoclonal Antibody [clone: 5E805]

Fornitore: United States Biological
USBIT5300-10DEA 526 CHF
USBIT5300-10D
Anti-TG Mouse Monoclonal Antibody [clone: 5E805]
Anticorpi
Type: Primary
Antigen: TG (thyroglobulin)
Clonality: monoclonal
Clone: 5E805
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine, Cat, Dog, Human, Mouse, Pig, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR