Stampa…

Anti-TG Mouse Monoclonal Antibody [clone: 10F527]

Fornitore: United States Biological
USBIT5300-02EEA 691 CHF
USBIT5300-02E
Anti-TG Mouse Monoclonal Antibody [clone: 10F527]
Anticorpi
Type: Primary
Antigen: TG (thyroglobulin)
Clonality: monoclonal
Clone: 10F527
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR