Stampa…

Anti-TYMS Rabbit Monoclonal Antibody [clone: D26G11]

Fornitore: United States Biological
USBIT5155-10KEA 739 CHF
USBIT5155-10K
Anti-TYMS Rabbit Monoclonal Antibody [clone: D26G11]
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100

Type: Primary
Antigen: TYMS (thymidylate synthetase)
Clonality: monoclonal
Clone: D26G11
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Monkey, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR