Stampa…

Anti-Thymic Stromal Lymphopoietin Protein Receptor Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5150-07EA 739 CHF
USBIT5150-07
Anti-Thymic Stromal Lymphopoietin Protein Receptor Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Thymic Stromal Lymphopoietin Protein Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR