Stampa…

Anti-Thymic Factor Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5150-01B-20EA 502 CHF
USBIT5150-01B-20
Anti-Thymic Factor Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:10000-1:15000

Type: Primary
Antigen: Thymic Factor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR