Stampa…

Anti-THRSP Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT5051-10EA 889 CHF
USBIT5051-10
Anti-THRSP Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: THRSP (thyroid hormone responsive)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR