Stampa…

Anti-THPO Sheep Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT5050-09AEA 1180 CHF
USBIT5050-09A
Anti-THPO Sheep Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: THPO (thrombopoietin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR