Stampa…
Anti-THPO Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT5050-06REA 891 CHF
USBIT5050-06R
Anti-THPO Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:800; Immunochemistry: 1:500
Type: Primary
Antigen: THPO (thrombopoietin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR