Stampa…

Anti-THPO Mouse Monoclonal Antibody [clone: 7C200]

Fornitore: United States Biological
USBIT5050-06NEA 603 CHF
USBIT5050-06N
Anti-THPO Mouse Monoclonal Antibody [clone: 7C200]
Anticorpi
Recommended dilution: ELISA: 1:4000; Western Blot: 1:1000

Type: Primary
Antigen: THPO (thrombopoietin)
Clonality: monoclonal
Clone: 7C200
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR