Stampa…

Anti-THPO Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT5050-06KEA 922 CHF
USBIT5050-06K
Anti-THPO Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: THPO (thrombopoietin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR