Stampa…

Anti-TXN Rabbit Monoclonal Antibody [clone: 7H76]

Fornitore: United States Biological
USBIT5010-10HEA 739 CHF
USBIT5010-10H
Anti-TXN Rabbit Monoclonal Antibody [clone: 7H76]
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:400

Type: Primary
Antigen: TXN (thioredoxin)
Clonality: monoclonal
Clone: 7H76
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR