Stampa…

Anti-Thioredoxin-1 Mouse Monoclonal Antibody [clone: 10K365]

Fornitore: United States Biological
USBIT5010-09DEA 672 CHF
USBIT5010-09D
Anti-Thioredoxin-1 Mouse Monoclonal Antibody [clone: 10K365]
Anticorpi
Recommended dilution: Western Blot: 1:2000

Type: Primary
Antigen: Thioredoxin-1
Clonality: monoclonal
Clone: 10K365
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR