Stampa…

Anti-ACOT13 Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT3773-04-BIOTEA 1200 CHF
USBIT3773-04-BIOT
Anti-ACOT13 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: ACOT13 (acyl-CoA thioesterase 13)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR