Stampa…

Anti-THAP11 Mouse Monoclonal Antibody [clone: 11D112]

Fornitore: United States Biological
USBIT3700-02EA 1050 CHF
USBIT3700-02
Anti-THAP11 Mouse Monoclonal Antibody [clone: 11D112]
Anticorpi
Type: Primary
Antigen: THAP11 (THAP domain containing 11)
Clonality: monoclonal
Clone: 11D112
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR