Stampa…
Anti-TGOLN2 Sheep Polyclonal Antibody
Fornitore: United States Biological
USBIT3660-08AEA 1280 CHF
USBIT3660-08A
Anti-TGOLN2 Sheep Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 1:100-1:200; Western Blot: 1:1000
Type: Primary
Antigen: TGOLN2 (trans-golgi network protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat
Mehr About VWR