Stampa…

Anti-GTF2I Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT3599-05HEA 825 CHF
USBIT3599-05H
Anti-GTF2I Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: GTF2I (general transcription factor IIi)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR