Stampa…

Anti-TFB2M Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT3597-75CEA 774 CHF
USBIT3597-75C
Anti-TFB2M Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:128000

Type: Primary
Antigen: TFB2M (transcription factor B2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR