Stampa…

Anti-TTC5 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT3503-15EA 739 CHF
USBIT3503-15
Anti-TTC5 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 05-1 ?g/ml

Type: Primary
Antigen: TTC5 (tetratricopeptide repeat domain 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR