Stampa…

Anti-TTC1 Mouse Monoclonal Antibody [clone: 4E3]

Fornitore: United States Biological
USBIT3502-90EA 988 CHF
USBIT3502-90
Anti-TTC1 Mouse Monoclonal Antibody [clone: 4E3]
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TTC1 (tetratricopeptide repeat domain 1)
Clonality: monoclonal
Clone: 4000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR