Stampa…

Anti-TSPAN31 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: United States Biological
USBIT3300-31-HRPEA 1200 CHF
USBIT3300-31-HRP
Anti-TSPAN31 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: TSPAN31 (tetraspanin 31)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR