Stampa…

Anti-TSPAN31 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornitore: United States Biological
USBIT3300-31-APEA 1200 CHF
USBIT3300-31-AP
Anti-TSPAN31 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: TSPAN31 (tetraspanin 31)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR