Stampa…
Anti-TSPAN12 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT3300-12EA 825 CHF
USBIT3300-12
Anti-TSPAN12 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: TSPAN12 (tetraspanin 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR