Stampa…
Anti-Testin Isoform 1 Chicken Polyclonal Antibody
Fornitore: United States Biological
USBIT2943-01EA 667 CHF
USBIT2943-01
Anti-Testin Isoform 1 Chicken Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:250; Immunochemistry: 1:100;
Type: Primary
Antigen: Testin Isoform 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Mehr About VWR