Stampa…

Anti-Terminal Deoxynucleotidyl Transferase Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT2750-04EA 761 CHF
USBIT2750-04
Anti-Terminal Deoxynucleotidyl Transferase Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: Terminal Deoxynucleotidyl Transferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR