Stampa…
Anti-Telomerase Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT2399-70-100EA 966 CHF
USBIT2399-70-100
Anti-Telomerase Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Telomerase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Mehr About VWR